Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosine Hydroxylase Rabbit anti-Human, Rat, Mouse, Feline, Ferret, Squid, Drosophila, Mollusk, Polyclonal, MilliporeSigma™
GSA_VA

Catalog No. AB152MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB152MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB152MI Supplier MilliporeSigma Supplier No. AB152
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Anti-Tyrosine Hydroxylase Antibody detects level of TH and has been published and validated for use in ELISA, immunofluoresence, immunohistochemistry, immunohistochemistry (paraffin), immunoprecipitation and western blotting.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Tyrosine Hydroxylase
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing liquid in 10mM HEPES (pH 7.5), 150mM NaCl (pH 7.5), 100 ug/mL BSA and 50% glycerol.
Gene Symbols TH; TYH; EC 1.14.16.2
Host Species Rabbit
Immunogen Denatured tyrosine hydroxylase from rat pheochromocytoma (denatured by sodium dodecyl sulfate).
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_012740.3
Target Species Drosophila, Feline, Ferret, Human, Mollusca, Mouse, Rat, Squid
Content And Storage −20-−70°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.