Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosine Hydroxylase Mouse anti-Chicken, Frog, Human, Mouse, Monkey, Rat, Lizard, Vole, Zebrafish, Clone: LNC1, MilliporeSigma™

Catalog No. MAB318MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB318MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB318MI Supplier MilliporeSigma Supplier No. MAB318
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Tyrosine Hydroxylase using this Anti-Tyrosine Hydroxylase Antibody, clone LNC1 validated for use in immunohistochemistry, immunohistochemistry (paraffin), immunoprecipitation and western blotting with more than 85 product citations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tyrosine Hydroxylase
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LNC1
Conjugate Unconjugated
Formulation Ascites mouse monoclonal IgG1κ fluid containing 3% BSA. Contains no preservative.
Gene Accession No. NM_000360.3
Gene Symbols TH, TYH, EC 1.14.16.2
Host Species Mouse
Immunogen Tyrosine Hydroxylase purified from PC12 cells
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Chicken, Frog, Human, Lizard, Monkey, Mouse, Rat, Zebrafish
Content And Storage −20°C for up to 12 months.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.