Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004358 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-358 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-358 Supplier Abnova Corporation Supplier No. H00007322A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant UBE2D2.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme functions in the ubiquitination of the tumor-suppressor protein p53, which is induced by an E3 ubiquitin-protein ligase. Two alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms. [provided by RefSeq

Sequence: MALKRIHKELNDLARDPPAQCSAGPVGDDMFHWQATIMGPNDSPYQGGVFFLTIHFPTDYPFKPPKVAFTTRIYHPNINSNGSICLDILRSQWS

Specifications

Antigen ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant UBE2D2.
Formulation 50% glycerol
Gene UBE2D2
Gene Accession No. NM_003339
Gene Alias E2(17)KB2/PUBC1/UBC4/UBC4/5/UBCH5B
Gene Symbols UBE2D2
Host Species Mouse
Immunogen UBE2D2 (NP_003330, 1 a.a. ∼ 94 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7322
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.