Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vβ 14 TCR Rat anti-Mouse, FITC, Clone: 14-2, BD

Catalog No. BDB553258 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553258 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553258 Supplier BD Biosciences Supplier No. 553258
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 14-2 antibody reacts with the Vβ 14 T-Cell Receptor (TCR) of mice having the a (e.g., C57BR, C57L, SJL, SWR), b (e.g., A, AKR, BALB/c, CBA, C3H/He, C57BL, C58, DBA/1, DBA/2), and c (e.g., RIII) halpotypes of the Tcrb gene complex. Vβ 14 TCR-

Host Species: Rat
Clone: 14-2
Isotype: IgM, κ
Species Reactivity: Mouse
Immunogen: Mouse B10.A Helper T-Cell Clone J9.19

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vβ 14 TCR
Applications Flow Cytometry
Classification Monoclonal
Clone 14-2
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse B10.A Helper T-Cell Clone J9.19
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.