Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-v-H-Ras Clone: Y13-259,

Catalog No. OP01100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP01100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP01100UG Supplier MilliporeSigma™ Supplier No. OP01100UG
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects v-H-Ras Clone: Y13-259 in Human, Mouse, Rat samples, and it is validated for Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Immuno Blotting

Oncogene sequences originally isolated from Harvey sarcoma virus v-H-ras and Kirsten sarcoma virus v-K-ras, both encode 21,000 dalton phosphoproteins (p21's) with guanine nucleotide binding activity. Human cellular homologues of v-H-ras and v-K-ras, designated c-H-ras and c-K-ras, respectively, and a related human oncogene, c-N-ras, each encode 21,000 dalton protein analogues to the gene products of their viral counterparts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen v-H-Ras
Applications Immunoblot, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization
Classification Monoclonal
Clone Y13-259
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin.
Host Species Rat
Immunogen recombinant p21 Ras
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes both phosphorylated and nonphosphorylated forms of v-H-Ras, and v-K-Ras in SW480 and Y1 cells and normal skin tissue. Also recognizes H, K, and N-Ras human translated products.
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.