Learn More
Vinculin Mouse anti-Chicken, Human, Mouse, Rat, Rabbit, Clone: V284, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Vinculin |
| Applications | Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | V284 |
| Conjugate | Unconjugated |
| Formulation | PBS, pH 7.4, containing 0.05% sodium azide and 1mg/ml BSA |
| Gene Accession No. | NM_003373 |
| Gene Symbols | VCL, Metavinculin, OTTHUMP00000043885, vinculin, MVCL, Vinculin |
| Host Species | Mouse |
| Immunogen | Human Vinculin |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.