Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WASH complex subunit FAM21C Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABT79MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT79MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT79MI Supplier MilliporeSigma Supplier No. ABT79
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-WASH complex subunit FAM21C Antibody detects level of WASH complex subunit FAM21C and has been published and validated for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WASH complex subunit FAM21C
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols FAM21C; FAM21A; VPEF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to human WASH complex subunit FAM21C.
Purification Method Protein G Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_056077
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.