Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XBP-1S Mouse, Brilliant Violet 421, Clone: Q3-695, BD

Catalog No. BDB563382 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563382 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563382 Supplier BD Biosciences Supplier No. 563382
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Q3-695 monoclonal antibody specifically binds to XBP-1S, the spliced isoform of X-box binding protein 1 (XBP-1). XBP-1S is generated by cells that undergo an unfolded protein response, ie, cells responding to the accumulation of unfolded proteins in the endoplasmic reticulum. It plays important roles as a transcription factor in a number of important cellular processes including the regulation of MHC class II genes, differentiation of plasma cells, immunoglobulin secretion and hepatocyte growth.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen XBP-1S
Applications Flow Cytometry
Classification Monoclonal
Clone Q3-695
Conjugate Brilliant Violet 421
Description XBP1; X-box binding protein 1; TREB5; Tax-responsive element-binding
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human XBP-1S Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.