Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAP-70 (pY319)/Syk (pY352) Mouse anti-Human, Unlabeled, Clone: 17A/P-ZAP70, BD

Catalog No. BDB612574 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612574 50 μg
BDB612575 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612574 Supplier BD Biosciences Supplier No. 612574
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZAP70 is a protein tyrosine kinase (PTK) that associates with the z subunit of the T cell antigen receptor (TCR) and undergoes tyrosine phosphorylation following TCR stimulation. ZAP70 contains two SH2-like domains with the PTK domain located at the C-terminus. It appears that both ZAP70 and Syk are recruited to the phosphorylated CD3 and z subunits after TCR stimulation. TCR stimulation leads to autophosphorylation of ZAP70 at Tyr-315 amd Tyr-319, and mutation of the Tyr-319 site dramatically impairs TCR signaling. In addition, TCR-mediated Lck activity leads to phosphorylation of ZAP70 on Tyr-493 in the regulatory loop of the kinase domain leading to upregulation of ZAP70 kinase activity. The significance of ZAP70 activation in mediating TCR signal transduction has been confirmed by showing that ZAP70 activity is absent in an autosomal recessive form of severe combined immunodeficiency (SCID). This is due to mutations affecting the ZAP70 kinase domain which affect the stability of the protein and TCR signaling. Clone 17A/P-ZAP70 recognizes the phosphorylated form of ZAP70 (Y319). It also cross-reacts with SYK (Y352) due to homology of the phosphorylation site with ZAP70 (Y319). The PE-conjugated format has been evaluated using human and mouse model systems. The unconjugated form of the antibody has also been shown to work in western blot analysis on human, mouse, and rat cells.

Flow Cytometry, Intracellular Staining, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZAP-70 (pY319)/Syk (pY352)
Applications Western Blot
Classification Monoclonal
Clone 17A/P-ZAP70
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human phosphorylated ZAP70 Peptide
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.