Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZO-1 Mouse anti-Human, Unlabeled, Clone: 1, BD

Catalog No. p-3852033 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610966 50 μg
BDB610967 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610966 Supplier BD Biosciences Supplier No. 610966
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tight junctions ( z onulae o ccludens) are critical to the maintenance of cell polarity and intracellular barriers between epithelial and endothelial cells. Protein components of the tight junctions include actin filaments, symplekin, occludin, Rab3B, AF-6, 7H6, ZO-1, and ZO-2. Analysis of ZO-1 and -2 resulted in their inclusion in the MAGUK protein family. This family also includes the discs large tumor suppressor protein (Dlg-A) of Drosophila ; p55, an erythrocyte membrane protein; and PSD-95/SAP90, a synaptic membrane protein. All family members contain a region homologous to guanylate kinase (GuK), a src homology (SH3) domain, and multiple PDZ domains. Through these elements, MAGUK proteins function in signal transduction and, possibly, tumor suppression. ZO-1 is a peripheral membrane phosphoprotein that binds to other tight junction proteins such as occludin and AF-6. Via its SH3 domain, ZO-1 interacts with a serine protein kinase that phosphorylates a region immediately C-terminal of the SH3 domain. Taken together, these data indicate that ZO-1 is a critical element in the formation of tight junctions and may also serve an important role in signaling and tumor suppression.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZO-1
Applications Western Blot
Classification Monoclonal
Clone 1
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human ZO-1 aa. 1048-1247
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.