Learn More
Description
- Application of Advanced Plasma Treatment technology (APT) allows for comprehensive complete hydrophilic treatment that is not compromised in presence of solvents
- Meets the SBS and ANSI dimensions used for automation
- Conical shaped bottoms for maximum sample recovery and efficient mixing
- Manufactured from polypropylene resin
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | Natural |
| Material | Polypropylene |
| No. of Wells | 96 |
| Surface Treatment | AntiBIND |
| Total Volume (Metric) Well | 0.5 mL |
| Well Shape | Conical Bottom (V-well) |
| For Use With (Application) | High through-put drug discovery, proteomics research, bioanalytical assays, long term sample storage, sample prep, chromatography (LC-MS) |
| No. per Case | 120 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
