Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ AntiBIND™ Deep Well Microplates
SDP

Catalog No. 02912402 Shop All DWK Life Sciences Products
Change view
Click to view available options
Total Volume (Metric) Well:
0.5 mL
1 mL
No. per Case:
40
120
No. of Wells:
96
Array:
8 x 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Total Volume (Metric) Well No. per Case No. of Wells Array
02-912-402 1 mL 40 96 8 x 12
02-911-702 0.5 mL 120 96 8 x 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-912-402 Supplier DWK Life Sciences Supplier No. W266028

missing translation for 'validMainframeMsg'

Used in chromatography applications, long term sample storage, and bioanalytical assays. Wheaton AntiBIND 96 Deep Well Microplates, Conical, are manufactured using a new technology that modifies the polypropylene surface at the molecular level to produce a homogenous hydrophilic surface that eliminates nonspecific protein binding and ensures ultimate recovery of valuable proteins.

  • Application of Advanced Plasma Treatment technology (APT) allows for comprehensive complete hydrophilic treatment that is not compromised in presence of solvents
  • Meets the SBS and ANSI dimensions used for automation
  • Conical shaped bottoms for maximum sample recovery and efficient mixing
  • Manufactured from polypropylene resin
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Color Natural
Product Type Deep-Well Microplate
Volume (Metric) Well 1 mL
No. of Wells 96
Surface Treatment AntiBIND
Total Volume (Metric) Well 1 mL
Well Shape Conical Bottom (V-well)
For Use With (Application) High through-put drug discovery, proteomics research, bioanalytical assays, long term sample storage, sample prep, chromatography (LC-MS)
No. per Case 40
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.