Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aldon™ Antibiotic Disks
SCI_ED

Catalog No. S28509 Shop All Aldon Products
Change view
Click to view available options
Product Type:
Ampicillin disks
Chloramphenicol Disks
Gentamicin Disks
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S28509 Gentamicin Disks
S28507 Ampicillin disks
S28508 Chloramphenicol Disks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S28509 Supplier Aldon™ Supplier No. XD1363
Add to Cart
Add to Cart

Determines antibiotic susceptibility of microorganisms.

  • Each cartridge contains 50 disks (10mcg) of antibiotics with reclosable cap
  • Disks are 6.35mm (0.25 in.) in diameter
  • DOT Info: Non-regulated

Specifications

Grade 7 to 14
Format Disk
Product Type Gentamicin Disks
Includes Gentamicin disks
Diameter (English) Ring 0.25 in.
Quantity 50 Disks of 10 mcg
Color White
Languages English
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.