Learn More
Abcam Antibiotic PF 1052, Neutrophil Migration Inhibitor, Solid, 1 mg
Description
- Molecular weight: 429.59 g/mol
- Purity: >98%
- Physical form: Solid
- Activity: Neutrophil migration inhibitor; shows antibacterial and potentially antifungal and phytotoxic properties; active in vivo and in vitro
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | Neutrophil |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Quantity | 1 mg |
| For Use With (Application) | Neutrophil migration inhibition, Antibacterial research, Antifungal research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.