Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Antibiotic PF 1052, Neutrophil Migration Inhibitor, Solid, 500 μg

Catalog No. 502706005
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6005 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6005 Supplier Abcam Supplier No. AB144183500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Potent neutrophil migration inhibitor with antibacterial, antifungal, and phytotoxic properties.

  • Molecular Weight: 429.59 g/mol
  • Purity: >98%
  • Form: Solid
  • Activity: Neutrophil migration inhibitor
  • Properties: Antibacterial, potentially antifungal and phytotoxic properties
  • Activity context: Active in vivo and in vitro
  • CAS Number: 147317-15-5

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Target Molecule Neutrophil
Substrate Properties Chemical Substrate
Buffer 100% Antibiotic PF 1052
Shipping Condition Ambient
Form Solid
Quantity 500 μg
For Use With (Application) Neutrophil migration inhibition, Antibacterial, Antifungal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.