Learn More
Antibiotic Sensitivity Discs
Study antibiotic sensitivity and resistance.
Supplier: Fischer Technical Company B31328
This item is not returnable.
View return policy
Description
- Vials are sealed for storage
- Designed to dispense one disc at a time, without contact or spoilage of remaining discs
Order Info
Vials and bottles are supplied in packs of 50.
Specifications
Streptomycin | |
10μg | |
Disc | |
50 Discs |