Learn More
Cayman Chemical ANTIBODY 1-BCP 25mg
SDP
Supplier:  Cayman Chemical 3342825 MG
Encompass_Preferred
A dammarane-type triterpene saponin prevents increases in iNOS IL-6 and TNF-a levels as well as phosphorylation of NF-KB IKB p38 MAPK and Akt induced by LPS and TNF-a in rat C6 glial at 25 M reduces LPS-induced increases in NF-KB IKB p38 MAPK and Akt phosphorylation in mouse brain cortex at 30 mg/kg per day
Catalog No. 50-294-9731
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.