Learn More
Cayman Chemical ANTIBODY 1-BCP 5mg
SDP
Supplier:  Cayman Chemical 334285 MG
Encompass_Preferred
A prodrug form of niflumic acid decreases the rate of chloride/bicarbonate exchange in isolated mouse intestinal villous epithelial cells at 100 UM dietary administration increases survival but has no effect on the number of crypt goblet cells in the jejunum or cecum in a cystic fibrosis mouse model of distal intestinal obstructive syndrome at 88 mg/kg
Catalog No. 50-294-9976
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.