Learn More
Cayman Chemical ANTIBODY 15-OxoETE 250ug
SDP
Supplier:  Cayman Chemical 34730250 UG
Encompass_Preferred
A positive allosteric modulator of mGluR5 receptors potentiates glutamate-induced calcium mobilization in MCB3901 cells expressing human mGluR5 (EC50 52 nM) increases rat hippocampal mGluR5 receptor occupancy (ED50 13 mg/kg) increases wakefulness and decreases NREM and REM sleep in rats at 0 3 1 and 3 mg/kg
Catalog No. 50-291-0253
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.