Learn More
ACROBiosystems Monoclonal Anti-DNP antibody, Human IgG4 Isotype Control
Supplier:  ACROBiosystems DNPM31MG
Encompass_Preferred
Antibody;1MG;Human IgG4/kappa;Mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with DNP F(ab')2.; Dinitrophenyl (DNP) is a hapten that is not normally expressed in the target tissue. Therefore, anti-DNP antibody has no relevant specificity to a target antigen, and can be used as isotype control antibody to differentiate non-specific background signal from specific antibody signal in various in vitro and in vivo studies.Monoclonal Anti-DNP (Anti-Hapten) antibody, Human IgG4 was purified from HEK293 cell culture at ACRObiosystems, and specifically reacts with DNP (Dinitrophenyl) and DNP conjugated proteins.
Catalog No. 50-201-9864
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.