Learn More
Cayman Chemical ANTIBODY 4-IPP 10mg
SDP
Supplier:  Cayman Chemical 3177010 MG
Encompass_Preferred
An adenosine A3 receptor agonist binds selectively to adenosine A3 receptors (Ki 0 33 nM) over A1 and A2A receptors (Kis 820 and 470 nM respectively) and inhibits adenylate cyclase activity (IC50 66 8 nM) inhibits proliferation of B16/F10 melanoma cells and increases proliferation of murine bone marrow cells in vitro at 10 and 100 nM induces accumulation of cells in the G0/G1 phase of the cell cycle and induces apoptosis at 100 nM prevents the development of lung metastatic foci in a B16/F10 mouse model of lung metastasis and reverses decreases in the number of white blood cells and neutrophils induced by cyclophosphamide at 6 Ug/kg
Catalog No. 50-299-0884
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.