Learn More
Cayman Chemical ANTIBODY 5-BDBD 10mg
SDP
Supplier:  Cayman Chemical 4203410 MG
Encompass_Preferred
An isoxazoline ectoparasiticide It inhibits GABA-induced chloride currents in CHO-K1 cells expressing either CfRDL-A285 or CfRDL-S285 (IC50s 135 and 136 nM respectively) induces mortality in C. felis fleas (LC80 0.3 Ug/ml) and O. turicata soft ticks (LC100 0.003 Ug/ml) eliminates live C. felis fleas as well as R. sanguineus I. ricinus and D. reticulatus ticks on dogs after 48 hours and reduces the number of fleas and ticks for at least 35 days at 2.5 mg/kg
Catalog No. 50-294-5633
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.