Learn More
Cayman Chemical ANTIBODY 5S 6R-EET 50UG
SDP
Supplier:  Cayman Chemical 900434950 UG
Encompass_Preferred
A naphthoquinone that has diverse biological activities inhibits a panel of 10 CYPs (IC50s 1.4-4.0 UM) cytotoxic to H1975 A549 PC-9 and H460 NSCLC cells (IC50s 3.9 4.7 20.7 and 17.3 UM respectively) inhibits the migration of as well as reduces EGFR and STAT3 signaling and induces apoptosis in H1975 PC-9 and H460 cells at 0.5 1 and 2 UM reduces tumor weight and intratumoral STAT3 phosphorylation in an H1975 mouse xenograft model at 1 mg/kg inhibits SARS-CoV-2 PLpro (IC50 5.05 UM) reduces BALF levels of Tnf-a Il-1a and Il-6 in a mouse model of acute lung injury induced by poly (IC) and SARS-CoV-2 spike glycoprotein at 300 mg/kg
Catalog No. 50-289-5801
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.