Learn More
Cayman Chemical ANTIBODY AQC 250mg
SDP
Supplier:  Cayman Chemical 30877250 MG
Encompass_Preferred
A form of tocopherol has similar antioxidant activity to a-tocol but lower activity than Y-tocol increases microviscosity of PC rat liver liposomes by 70 6 when used at a molar ratio of 0 2 to PC has been used as an internal standard for the quantification of tocols tocopherols and tocopheryl quinones by HPLC
Catalog No. 50-299-0274
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.