Learn More
Cayman Chemical ANTIBODY AZ-GHS-22 1mg
SDP
Supplier:  Cayman Chemical 346111 MG
Encompass_Preferred
A histamine H2 receptor antagonist with gastroprotective activity inhibits histamine-induced cAMP production in CHO cells expressing human histamine H2 receptors at 10 nM reduces hemorrhagic esophageal lesion size and gastric acid secretion in a rat model of pyloric ligation-induced reflux esophagitis at 3 10 and 30 mg/kg prevents 5-fluorouracil-induced intestinal mucositis diarrhea and body weight loss in wild-type but not Trpv1-/- or sensory deafferented mice from 3-30 mg/kg reduces indomethacin-induced antral ulcer size in wild-type but not chemically-deafferented rats at 10 mg/kg
Catalog No. 50-294-8417
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.