Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Antibody Column Package: Modern Protein A (1-step Purification Protocol)

Catalog No. 09929010 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-929-010 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-929-010 Supplier Cytiva Supplier No. 29497628
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Combines affinity chromatography with desalting for the purification of human antibodies

  • Contains columns for a 1-step protocol using affinity chromatography: HiTrap™ MabSelect™ PrismA 1 × 1 mL and HiTrap™ Desalting 1 × 5 mL
  • HiTrap™ MabSelect PrismA column is prepacked with MabSelect PrismA resin for preparative purification of monoclonal and polyclonal antibodies from serum and cell culture supernatant
  • HiTrap™ Desalting column is packed with Sephadex G-25 to give quick pH neutralization of the eluted antibodies (0.1 to 1.5 mL is the recommended sample loading volume)
  • Includes instructions with guidance and tips to quickly get started
TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Column Pack
Format Prepacked Column
Ligand Protein A; Protein A, Alkali Stabilized
For Use With (Application) Purification of Antibodies
Description Resin Type: MabSelect
Content And Storage HiTrap™ MabSelect™ PrismA (1 x 1 mL), and HiTrap™ Desalting (1 x 5 mL). 2°C to 8°C
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.