Learn More
Cytiva Antibody Column Package: Modern Protein A (2-step Purification Protocol)
Shop All Cytiva ProductsDescription
- Contains columns suitable for a successful 2-step protocol of human antibodies: HiTrap™ MabSelect™ PrismA (1 × 1 mL) and HiPrep™ 16/60 Sephacryl S-300 HR
- Includes instructions with guidance and tips to quickly get started
- HiTrap™ MabSelect PrismA column is prepacked with MabSelect PrismA resin for preparative purification of monoclonal and polyclonal antibodies from serum and cell culture supernatant, providing excellent purity in one step
- A final high purity is obtained with the HiPrep 16/60 Sephacryl S-300 HR size exclusion column, which removes antibody aggregates and/or fragments
Specifications
Specifications
| Product Type | Antibody Column Pack |
| Format | Prepacked Column |
| Ligand | Protein A; Protein A, Alkali Stabilized |
| For Use With (Application) | Purification of Antibodies |
| Description | Resin Type: MabSelect |
| Content And Storage | HiTrap™ MabSelect™ PrismA (1 x 1 mL), and HiPrep™ 16/60 Sephacryl S-300 HR Columns. 2°C to 8°C |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.