Learn More
Cayman Chemical ANTIBODY DavercIn 25mg
SDP
Supplier:  Cayman Chemical 2134825 MG
Encompass_Preferred
An orally bioavailable inhibitor of HDAC1 HDAC3 and HDAC6 (IC50s 43-72 nM) resulting in hyperacetylation of histone H4 in multiple myeloma cells abrogates cell growth and strongly induces apoptosis in multiple myeloma cells (IC50s 2 5-3 UM) dose-dependently inhibits HDAC activity in vivo
Catalog No. 50-298-5030
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.