Learn More
Abcam Antibody Diluent (HAMA Blocker) for ELISA, 50 mL
Catalog No. NC3074015
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
Catalog No. Quantity
NC3074015 50 mL
1 options
Catalog No. NC3074015 Supplier Abcam Supplier No. AB31812950ML
Abcam Antibody Diluent (HAMA Blocker) for ELISA.
Specifications
Product Type Diluent
Quantity 50 mL
For Use With (Application) ELISA