Learn More
Cayman Chemical ANTIBODY EnoxImon 10mg
SDP
Supplier:  Cayman Chemical 3086810 MG
Encompass_Preferred
A Notch inhibitor induces cell cycle arrest at the G0/G1 phase and decreases MYC and CCNA1 expression in Caki human renal cancer cells reduces tumor growth and increases survival in a 769-P mouse xenograft model at 8 mg/kg per day
Catalog No. 50-299-0397
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.