Learn More
Cayman Chemical ANTIBODY G150 1mg
SDP
Supplier:  Cayman Chemical 283181 MG
Encompass_Preferred
An inhibitor of topoisomerase I that inhibits topoisomerase I-mediated DNA cleavage 10-fold more potently than topotecan inhibits the growth of and is cytotoxic to human and mouse bone marrow cells in vitro (IC50s 0 4 and 2 3 nM respectively IC90s 1 2 and 8 4 nM respectively) inhibits the growth of and is cytotoxic to human tumor cell lines in vitro (IC50s 0 15-2 nM IC90s 0 7-8 3 nM) inhibits tumor growth in mouse xenograft models (1 36-2 7 mg/kg)
Catalog No. 50-294-0213
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.