Learn More
Cayman Chemical ANTIBODY GM 6001 1mg
SDP
Supplier:  Cayman Chemical 145331 MG
Encompass_Preferred
A RIPK1 inhibitor inhibits RIPK1 activity at 0.3 3 and 30 UM inhibits TNF-a-induced necroptosis in FADD-deficient Jurkat cells (EC50 180 nM) reduces infarct size when administered i.c.v. pre- and post-occlusion or post-occlusion only as 2 Ul of a 4 mM solution in a mouse model of MCAO
Catalog No. 50-296-9993
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.