Learn More
Cayman Chemical ANTIBODY GSK-A1 1mg
SDP
Supplier:  Cayman Chemical 345021 MG
Encompass_Preferred
A TRPA1 agonist (EC50 0.65 nM in a calcium influx assay) selective for TRPA1 over TRPV1 TRPV3 and TRPV4 TRPC5 TRPM2 and TRPM8 channels at 1 UM
Catalog No. 50-295-0706
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.