Learn More
BioVendor, LLC HEPATITIS B PROTEIN X VIRUS HUMAN RABBIT POLYCLONAL ANTIBODY
SDP
Supplier:  BioVendor, LLC RD981038100
Encompass_Preferred
TypePolyclonal AntibodyApplicationsWestern blotting, ImmunoprecipitationSource of AntigenE. coliHostsRabbitPreparationThe antibody was raised in rabbits by immunization with the recombinant HBx.Amino Acid SequenceThe immunization antigen (17 kDa) is a protein containing 153 AA of recombinant HBx and one extra AA, N-terminal methionin (highlighted).MAARVCCQLDPARDVLCLRPVGAESRGRPVSGPFGTLPSPSSSAVPADHGAHLSLRGLPVCAFSSAGPCALRFTSARRMETTVNAHQVLPKVLHKRTLGLSAMSTTDLEAYFKDCLFKDWEELGEEIRLKVFVLGGCRHKLVCSPAPCNFFTSASpecies ReactivityHumanPurification MethodImmunoaffinity chromatography on a column with immobilized recombinant Hepatitis B Protein X.Antibody Content0.05 mg (determined by BCA method, BSA was used as a standard)FormulationThe antibody is lyophilized in 0.05 M phosphate buffer, 0.1 M NaCl, pH 7.2. AZIDE FREE.
Catalog No. 50-894-096
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.