Learn More
Cayman Chemical ANTIBODY DFSX-2 100mg
SDP
Supplier:  Cayman Chemical 41981100 MG
Encompass_Preferred
A muscarinic acetylcholine receptor antagonist (Kis 7.55 22.6 1.42 8.86 and 2.63 nM respectively for recombinant human M1-M5 receptors) binds to isolated guinea pig bladders (Kb 0.813 nM) and inhibits ACh release from isolated rat bladders (IC50 0.747 nM) reduces urine volume in orally water-loaded rats from 10-300 Ug/kg
Catalog No. 50-294-3376
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.