Learn More
Cayman Chemical ANTIBODY IMP-1710 500ug
SDP
Supplier:  Cayman Chemical 31391500 UG
Encompass_Preferred
A phenazine pigment and virulence factor produced by P aeruginosa increases ROS production in RAW 264 7 cells in a concentration-dependent manner increases release of elastase and MPO by fMLP- and cytochalasin B-activated human neutrophils at 1 55 and 3 12 M respectively reduces ciliary beat frequency in isolated guinea pig tracheal rings resulting in ciliary dyskinesia by 1 hour at 10 50 and 100 M and ciliary stasis by 5 hours at 50 and 100 M reduces tracheal mucus velocity in anesthetized guinea pigs when administered as a 100 or 200 ng bolus dose active against the plant pathogenic fungi F graminearum P grisea A solani F oxysporium and S sclerotiorum (MICs 2-50 g/ml)
Catalog No. 50-299-0933
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.