Description
- Provides a qualitative visual test without specialized equipment.
- Uses Protein A/G strips to detect suitable colored antibody conjugates.
Specifications
Specifications
| Quantity | 30 Tests |
| Product Type | Antibody detection confirmation kit |
| Product Line | Novus |