Learn More
Cayman Chemical ANTIBODY LefamulIn 25mg
SDP
Supplier:  Cayman Chemical 3628525 MG
Encompass_Preferred
An adenosine A2B receptor antagonist (Ki 1 97 nM) selective for adenosine A2B receptors over adenosine A1 A2A and A3 receptors (Kis 403 503 and 570 nM respectively) increases 28-day survival as well as decreases peritoneal bacterial growth and plasma levels of IL-6 TNF-a and MIP-2 in a mouse model of CLP-induced sepsis from 0 5-10 mg/kg reduces disease severity in a mouse model of EAE at 1 mg/kg
Catalog No. 50-291-2937
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.