Learn More
Cayman Chemical ANTIBODY LonIcerIn 1mg
SDP
Supplier:  Cayman Chemical 251481 MG
Encompass_Preferred
A potent and selective inhibitor of GSK3 (IC50s 1.5 and 0.9 for human recombinant GSK3a and GSK2B respectively) has 10-fold selectivity for GSK3 against a panel of 40 kinases at a concentration of 20 M induces time-dependent stabilization ofBcatenin total protein and activates Wnt signaling in vitro has potent antiproliferative activity in BRAF wild-type/NRAS mutant and BRAF mutant melanoma cell lines (IC50s 6.0-11.8 nM) but has limited to no activity in non-melanoma cell lines (IC50s 430 to 20000 nM) knockdown ofBcatenin in A375 and M14 melanoma cells induces LY2090314 resistance in vitro reduces tumor volume in a murine A375 xenograft model as a single agent and synergizes with decabarzine for a greater antitumor effect
Catalog No. 50-293-4932
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.