Learn More
Cayman Chemical ANTIBODY LYG-202 1mg
SDP
Supplier:  Cayman Chemical 219761 MG
Encompass_Preferred
An inactive analog of the PHGDH inhibitors NCT-502 and NCT-503
Catalog No. 50-298-7071
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.