Learn More
Cayman Chemical ANTIBODY MALT 1mg
SDP
Supplier:  Cayman Chemical 265971 MG
Encompass_Preferred
An anticancer agent inhibits the growth of Calu-6 NSCLC and A-375 melanoma cell lines (EC50s 10 and 25 M respectively) induces arrests the cell cycle at the G2/M phase and induces apoptosis of Calu-6 cells in a concentration-dependent manner induces non-apoptotic growth arrest in HUVECs and inhibits VEGF- FGF- and EGF-induced endothelial cord formation in vitro (EC50s 47 103 and 34 nM respectively) reduces hemoglobin content and blood vessel density in a Matrigel plug model of neoangiogenesis reduces tumor growth in a dose-dependent manner in a Calu-6 mouse xenograft model
Catalog No. 50-293-4814
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.