Learn More
Cayman Chemical ANTIBODY MEDICA 16 5mg
SDP
Supplier:  Cayman Chemical 902905 MG
Encompass_Preferred
Kanamycin ELISA HRP Tracer has been tested and formulated to work exclusively with Cayman’s Kanamycin ELISA Kit (Item No. 502370)
Catalog No. 50-289-6067
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.