Learn More
Cayman Chemical ANTIBODY ML-SA1 10mg
SDP
Supplier:  Cayman Chemical 2995810 MG
Encompass_Preferred
A fungal metabolite cytotoxic to various human solid tumor cell lines including MCF-7 PC3 LNCap and DU145 cells (IC50s 1 3 0 65 1 5 and 1 2 UM respectively) active against C albicans
Catalog No. 50-298-9742
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.