Learn More
Cayman Chemical ANTIBODY MOZ-IN-3 1mg
SDP
Supplier:  Cayman Chemical 274021 MG
Encompass_Preferred
A saponin with diverse biological activities induces proliferation migration and tube formation of HUVECs in an mTOR-dependent manner induces cell cycle arrest at the S and G2/M phases and promotes apoptosis of SH-SY5Y cells at 45 M increases cGMP levels and induces relaxation of isolated precontracted rat mesenteric arteries promotes angiogenesis and decreases wound diameter in a mouse model of punched-hole ear injury at 0 25 2 5 and 25 mg/kg decreases tail bleeding time and increases thrombus weight in a rat tail bleeding assay at 1 25 mg/kg increases mRNA expression of collagen expression fibroblast proliferation and scar formation genes promotes neovascularization reduces monocyte infiltration and shortens wound closure time in a db/db mouse model of diabetic foot ulcers
Catalog No. 50-294-1405
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.