Learn More
Cayman Chemical ANTIBODY NK 252 1mg
SDP
Supplier:  Cayman Chemical 191311 MG
Encompass_Preferred
PGF3a is a COX product of EPA The biosynthesis of PGF3a from EPA was demonstrated in vitro in human and rabbit ocular tissues 1145 It has only 25 affinity at the ovine luteal FP receptor compared to PGF2a 2058
Catalog No. 50-298-1331
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.