Learn More
Cayman Chemical ANTIBODY NS 309 10mg
SDP
Supplier:  Cayman Chemical 1888910 MG
Encompass_Preferred
A synthetic CB structurally related to AB-CHMINACA in which the carboxamide-linked aminocarbonyl-2-methylpropyl side chain is substituted with an ester-linked 33-dimethylbutyl methyl ester intended for research and forensic applications
Catalog No. 50-298-2383
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.