Learn More
Cayman Chemical ANTIBODY NVS-MlT-1 5mg
SDP
Supplier:  Cayman Chemical 413185 MG
Encompass_Preferred
An inhibitor of the protein-protein interaction between Ras and Raf-1 inhibits the interaction between H-Ras and Raf-1 in a yeast two-hybrid assay at 30 UM inhibits Ras-induced activation of Raf-1 and MEK1 activity in HT-1080 cells at 20 UM reduces tumor volume in an SW620 mouse xenograft model at 100 and 600 mg/kg alone or in combination with paclitaxel
Catalog No. 50-294-2796
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.