Learn More
Cayman Chemical ANTIBODY ODQ 50mg
SDP
Supplier:  Cayman Chemical 8141050 MG
Encompass_Preferred
A microbial metabolite with diverse biological activities induces cytotoxicity in HepG2 and A549 cancer cells (IC50 ~15 UG/ml for both) induces cell cycle arrest at the G0 phase in HepG2 cells at 15 UG/ml reduces NO production in LPS-stimulated RAW 264.7 macrophages at 5 10 and 15 UG/ml increases lipolysis activity in IBMX- dexamethasone- and insulin-matured 3T3-L1 adipocytes at 10 UM reduces blood glucose and serum insulin levels in mice fed a high-fat diet at 5 mg/kg per day
Catalog No. 50-289-4831
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.