Learn More
Cayman Chemical ANTIBODY PeramIvIr 5mg
SDP
Supplier:  Cayman Chemical 237655 MG
Encompass_Preferred
An inducer of Hsp expression that increases Hsp70 Hsp22 Hsp27 Hsp90 and Hsp105 levels in various cells and tissues orally bioavailable and has diverse effects in vivo including hepatoprotective neuroprotective and antiulcerative effects
Catalog No. 50-299-4282
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.