Learn More
Cayman Chemical ANTIBODY PNU 282987 5mg
SDP
Supplier:  Cayman Chemical 174245 MG
Encompass_Preferred
A natural macrocyclic antibiotic that inhibits RNA polymerase with selectivity for Gram-positive bacteria over Gram-negative bacteria (IC50s 0.4 and 6 M respectively) has potent antibacterial activity against most Gram-positive bacteria and effectively targets the Gram-positive C. difficile (MIC 12 ng/ml)
Catalog No. 50-297-9654
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.